missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Jouberin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA565106
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: QVAMYSDLPFK SPIRDISYHP FENMVAFCAF GQNEPILLYI YDFHVAQQEA EMFKRYNGTF PLPGIHQSQD ALCTCPKLPH QGSFQIDEFV HTESSSTKMQ LVKQ Highest antigen sequence identity to the following orthologs - mouse 88%, rat 87%.
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
Especificaciones
| Jouberin | |
| Polyclonal | |
| Unconjugated | |
| AHI1 | |
| 1700015F03Rik; Abelson helper integration site 1; abelson helper integration site 1 protein; abelson helper integration site 1 protein homolog; Ahi1; AHI-1; contatins SH3 and WD40 domains; D10Bwg0629e; dJ71N10.1; JBTS3; jouberin; ORF1; RP1-32B1.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54806 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N157 | |
| AHI1 | |
| Recombinant Human Jouberin. Recombinant protein control fragment (Product #RP-94850). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido