missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: LPLSFVLDVL EGRAQHPGVL YVQKQCSNLP SELPQLLPDL ESHVPWASEA LGKMPDAVNF WLGEAAAVTS LHKDHYENLY CVVSGEKHFL FHPPSDRPFI PYELYTPATY QLTE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The jumonji domain-containing protein (JMJD) family is defined by the presence of the JmjC domain that is observed in several diverse species. While several JMJD proteins have been identified as being involved in chromatin regulation, histone demethylation and development, the function of JMJD7 has not been identified. JMJD7 was initially thought to be a novel splice form of the phospholipase PLA2G4B which is located downstream of JMJD7 as most tissues express a read-through transcript of the two genes.
Especificaciones
Especificaciones
| Antígeno | JMJD7 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | JMJD7 |
| N.º de referencia del gen | P0C870 |
| Alias de gen | Bifunctional peptidase and (3S)-lysyl hydroxylase JMJD7; hCG_37726; JmjC domain-containing protein 7; Jmjd7; jumonji domain containing 7; jumonji domain-containing protein 7; L-lysine (3S)-hydroxylase Jmjd7 |
| Símbolos de los genes | JMJD7 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
