missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: PNVSDSKHSI ANAKFLETAK KDSDQSWVSD VVKVDLTQSS VTNASSGNDH LNMEKEKYVS YISPLSAVSV MEDKLHKRSP PPETIKSKLN TSVDTHKIKS SPSPEVVKPK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
The jumonji domain containing 1C protein (JMJD1C) was initially discovered in silico, and later suggested to be a candidate gene for autism. Like the related proteins JMJD1A and JMJD1B, JMJD1C is a histone H3K9 demethylase implicated in the nuclear hormone receptor-based transcriptional regulation. JMJD1C mRNA is highly expressed in undifferentiated embryonic stem (ES) cells as well as pancreatic islet, diffuse-type gastric cancer, and other tissues and tumors. The JMJD1C gene promoter contain bHLH-, AP-1-, and POU5F1-binding sites, and as preferential expression of POU5F1 has been reported in ES cells, pancreatic islet, and diffuse-type gastric cancer, it has been suggested that POU5F1-mediated expression of JMJD1C reactivates previously silenced genes in ES cells and diffuse-type gastric cancer. At least three isoforms of JMJD1C are known to exist.
Especificaciones
Especificaciones
| Antígeno | JMJD1C |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | JMJD1C |
| N.º de referencia del gen | Q15652 |
| Alias de gen | 5430433L24Rik; D630035I23Rik; DKFZp761F0118; FLJ14374; Jhdm2c; JMJD1C; Jmjdic; jumonji domain containing 1C; jumonji domain-containing protein 1C; Kiaa1380; Pr2; Probable JmjC domain-containing histone demethylation protein 2C; RP11-10C13.2; thyroid hormone receptor interactor 8; thyroid receptor-interacting protein 8; TR-interacting protein 8; TRIP8; TRIP-8 |
| Símbolos de los genes | JMJD1C |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?