missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JLP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Marca:  Invitrogen™ PA559182

Código de producto. 15777041

  • 582.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Immunogen sequence: SGQVDKASLC GSMTSNSSAE TDSLLGGITV VGCSAEGVTG AATSPSTNGA SPVMDKPPEM EAENSEVDEN VPTAEEATEA TEGNAGSAED TVDISQTGVY TEHVFTDPLG VQIPEDLSPV YQSSN Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

JLP
Polyclonal
Unconjugated
SPAG9
3110018C07Rik; 4733401I23Rik; 4831406C20Rik; AW552012; Cancer/testis antigen 89; c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; CT89; HLC4; HLC6; HLC-6; HSS; Human lung cancer oncogene 6 protein; Jip4; JIP-4; JLP; JNK interacting protein; JNK/SAPK-associated protein; JNK/SAPK-associated protein 2; JNK-associated leucine-zipper protein; JNK-interacting leucine zipper; JNK-interacting protein 4; Jsap2; JSAP2a; Kiaa0516; lung cancer oncogene 4; Mapk8ip4; Max-binding protein; mitogen-activated protein kinase 8-interacting protein 4; PHET; PIG6; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; SPAG9; sperm associated antigen 9; sperm associated antigen 9 variant 5; sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1; syd1
Rabbit
Antigen affinity chromatography
RUO
9043
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O60271
SPAG9
Recombinant protein corresponding to Human JLP. Recombinant protein control fragment (Product #RP-89951).
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado