missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JIP3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 15577641
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15577641 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15577641 Proveedor Invitrogen™ N.º de proveedor PA536369

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 147 kDa. Purity is >95% by SDS-PAGE.

c-Jun NH2-terminal kinases (JNKs) are distant members of the MAP kinase family. JNK1 is activated by dual phosphorylation at a Thr-Pro-Tyr motif in response to ultraviolet (UV) light, and it functions to phosphorylate c-Jun at amino terminal serine regulatory sites Ser 63 and Ser 73, resulting in transcriptional activation. Two additional JNK family members, JNK2 and JNK3, have been identified. JIP-1 (for JNK interacting protein-1) has been identified as a cytoplasmic inhibitor of JNK that retains JNK in the cytoplasm, thereby inhibiting JNK-regulated gene expression. Evidence suggests that JNK1 and JNK2 bind to JIP-1 with greater affinity than to ATF-2 and c-Jun, which are targets of the JNK signaling pathway. JIP-1 contains an amino terminal JNK binding domain and a carboxy terminal SH3 domain. ATF-2 and c-Jun also contain the JNK binding domain and are thought to compete with JIP-1 for JNK binding. Multiple splice variants of JIP-1, including JIP-1b, JIP-1c (also designated islet-brain 1 or IB-1), JIP-2a, JIP-2b and JIP-3, have been identified in brain.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno JIP3
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.05% sodium azide; pH 7.2
génica MAPK8IP3
N.º de referencia del gen Q9UPT6
Alias de gen BB120594; c-Jun NH2-terminal kinase (JNK)/stress-activated protein kinase-associated protein 1; C-jun-amino-terminal kinase interacting protein 3; C-Jun-amino-terminal kinase-interacting protein 3; D17Wsu15e; homolog of Drosophila Sunday driver 2; JIP3; JIP-3; JNK MAP kinase scaffold protein 3; JNK/SAPK-associated protein 1; JNK/SAPK-associated protein 1c; JNK/SAPK-associated protein-1; JNK/stress-activated protein kinase-associated protein 1; JNK-interacting protein 3; JSAP1; JSAP1a; JSAP1b; JSAP1c; JSAP1d; JUN/SAPK-associated protein 1; KIAA1066; MAPK8IP3; mitogen activated protein kinase 8 interacting protein 3; mitogen-activated protein kinase 8 interacting protein 3; mitogen-activated protein kinase 8-interacting protein 3; mKIAA1066; Sunday driver 2; syd; SYD2
Símbolos de los genes MAPK8IP3
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to amino acids 615-664 of human JIP-3.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23162
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.