Learn More
Invitrogen™ JDP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5120335
Descripción
Positive test controls include: HeLa, Jurkat, HepG2, Rat testis. The target is usually found in the following locations: Nucleus. Immunogen sequence: MVAGWPATPP AMMPGQIPDP SVTTGSLPGL GPLTGLPSSA LTVEELKYAD IRNLGAMIAP LHFLEVKLGK RPQPVKSELD EEEERRKRRR EKNKVAAARC RNKKKERTEF LQRESERLEL MNAELKTQIE ELKQERQQLI LMLNRHRPTC IVRTDSVKTP ESEGNPLLEQ LEKK.
Component of the AP-1 transcription factor that represses transactivation mediated by the Jun family of proteins. Involved in a variety of transcriptional responses associated with AP-1 such as UV-induced apoptosis, cell differentiation, tumorigenesis and antitumogeneris. Can also function as a repressor by recruiting histone deacetylase 3/HDAC3 to the promoter region of JUN. May control transcription via direct regulation of the modification of histones and the assembly of chromatin.
Especificaciones
| JDP2 | |
| Polyclonal | |
| Unconjugated | |
| JDP2 | |
| c-Jun dimerization protein 2; Jdp2; Jdp-2; Jun dimerization protein 2; Jundm2; Jundp2; progesterone receptor co-activator; TIF | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116674, 122953 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.54 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q78E65, Q8WYK2 | |
| JDP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-174 of human JDP2 (NP_001128521.1). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.