missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JAM3 Polyclonal Antibody
GREENER_CHOICE

Código de producto. 10533755
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10533755 100 μg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10533755

Marca: Invitrogen™ 409000

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

JAM3 Polyclonal Antibody for Western Blot, ICC/IF

JAM3 is involved in tight junctions and represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. Unlike other proteins in this family, the this protein is unable to adhere to leukocyte cell lines and only forms weak homotypic interactions. The encoded protein is a member of the junctional adhesion molecule protein family and acts as a receptor for another member of this family.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno JAM3
Aplicaciones Western Blot, Immunocytochemistry
Clasificación Polyclonal
Concentración 0.25 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.1% sodium azide; pH 7.4
génica JAM3
N.º de referencia del gen Q9BX67, Q9D8B7
Alias de gen 1110002N23Rik; JAM-2; JAM3; JAM-3; JAMC; JAM-C; Jcam3; junction adhesion molecule 3; junction cell adhesion molecule 3; junction cell adhesion molecule C; junctional adhesion molecule 3; Junctional adhesion molecule C; sJAM-C; Soluble form of JAM-C; UNQ859/PRO1868
Símbolos de los genes JAM3
Especie del huésped Rabbit
Inmunógeno Synthetic peptide derived from an internal region of the mouse and rat JAM-C proteins, which differs from human, bovine, and dog by one conservative amino acid replacement.
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 83700, 83964
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.