missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA5 Monoclonal Antibody (SAM-1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen 132600
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
Especificaciones
| ITGA5 | |
| Monoclonal | |
| Unconjugated | |
| P08648 | |
| ITGA5 | |
| Purified monocytes. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| SAM-1 | |
| ITGA5 | |
| FNRA, CD49e, VLA5A, ITGA5 | |
| Mouse | |
| Purified | |
| RUO | |
| 3678 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido