missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Itacitinib, MedChemExpress
Descripción
Itacitinib (INCB039110) is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. Itacitinib shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2
Especificaciones
Especificaciones
| Nombre del producto químico o material | Itacitinib |
| CAS | 1334298-90-6 |
| Color | Light Yellow |
| Notas de grado de pureza | Research |
| Fórmula molecular | C26H23F4N9O |
| Cantidad | 5 mg |
| Sinónimo | INCB039110 |
| Información de solubilidad | DMSO : ≥ 30 mg/mL (54.20 mM) |
| SMILES | N#CCC1(N2N=CC(C3=C4C(NC=C4)=NC=N3)=C2)CN(C5CCN(C(C6=C(F)C(C(F)(F)F)=NC=C6)=O)CC5)C1 |
| Peso molecular (g/mol) | 553.51 |
| Mostrar más |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?