missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ISL2 Polyclonal Antibody

Descripción
Antibody detects endogenous levels of total ISL2.
ISL LIM Homeobox 2 (ISL2) is a gene located on chromosome 15q23 that encodes a transcription factor belonging to the LIM-homeodomain family. These transcription factors play critical roles in the regulation of gene expression during embryonic development, particularly in the differentiation and maintenance of neuronal and pancreatic cell lineages. ISL2 is specifically involved in the development of the nervous system, where it contributes to motor neuron differentiation and connectivity, as well as in the development of certain retinal structures. The protein functions by binding to specific DNA sequences, thereby influencing the transcription of target genes essential for cellular differentiation and identity. Although less studied than its paralog ISL1, ISL2's expression and roles in developmental pathways are crucial for proper organogenesis and tissue specification. Dysregulation of ISL2 has potential implications in developmental disorders and diseases involving neural and pancreatic tissues. Ongoing research aims to further elucidate its functions and potential as a therapeutic target in developmental biology and disease.
Especificaciones
Especificaciones
| Antígeno | ISL2 |
| Aplicaciones | Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | ISL2 |
| N.º de referencia del gen | P50480, Q96A47, Q9CXV0 |
| Alias de gen | 3110001N10Rik; Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL LIM homeobox 2; ISL2; Isl-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; islet-2 |
| Símbolos de los genes | ISL2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?