missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA112811
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control for this product is spleen, thymus or peripheral blood leukocytes.
Interferons (IFNs) are involved in a multitude of immune interactions during viral infections and play a major role in both the induction and regulation of innate and adaptive antiviral mechanisms. During infection, host-virus interactions signal downstream molecules such as transcription factors such as IFN regulatory factor-3 (IRF3) which can act to stimulate transcription of IFN-a/b genes. IRF7 has been shown to play a role in the transcriptional activation of virus-inducible cellular genes, including interferon beta chain genes. IRF7 play a major role in the innate immune pathway, interacting with the Toll-like receptor (TLR) adaptor proteins MyD88 and Tirp/TRAM and functioning as an intermediate TLR4 and TLR9 signaling. There are at least four differentially spliced isoforms of IRF7, although their function has not been clearly established.
Especificaciones
| IRF7 | |
| Polyclonal | |
| Unconjugated | |
| IRF7 | |
| IRF-7H, IRF7A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3665 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q92985 | |
| IRF7 | |
| Peptide corresponding to 14 residues near the center of human IRF7. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido