missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Mouse anti-Human, Clone: OTI2G3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies TA500520
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IRF-3 is encoded by the IRF-3 gene located on chromosome 19 in humans and belongs to the Interferon regulatory transcription factor (IRF) family. IRF-3 plays a critical role in the response of the immune system to viral infection. It binds to the Interferon stimulated response element (ISRE) of its downstream target genes such as Interferon alpha and beta to activate their transcription which eventually leads to immune modulatory actions of the type I Interferon (IFN) pathway. Following phosphorylation on Serine 396, IRF-3 has been shown to affiliate with CREB binding protein (CREBBP) and translocate to the nucleus which is critical for its ability to bind to DNA and act as a transcription factor. Additionally, phosphorylation associated nuclear translocation may also protect IRF-3 from degradation by the virus machinery.Especificaciones
| IRF3 | |
| Monoclonal | |
| Unconjugated | |
| IRF3 | |
| IIAE7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3661 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G3 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14653 | |
| IRF3 | |
| Full-length protein expressed in 293T cell transfected with human IRF3 expression vector | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido