Learn More
IRF2BP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-93674
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IRF2BP2 Polyclonal specifically detects IRF2BP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Especificaciones
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| IRF2BP2 | |
| 0.1 mL |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QDWVNRPKTVRDTLLALHQHGHSGPFESKFKKEPALTAGRLLGFEANGANGSKAVARTARKRKPSPEPEGEVG | |
| Mouse, Rat |