Learn More
IRE1 alpha, p Ser724 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 252 publications
Marca: Novus Biologicals NB100-2323
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammal, Monkey, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In vitro assay, Knockdown Validated.
Especificaciones
| IRE1 alpha (p Ser724) | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, ire1 alpha p Ser724, ire1 alpha phosphorylation, IRE1a, ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, phospho ire1 alpha, phospho-ire1 alpha, phospho-ire1a, p-ire1 alpha, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| Rabbit | |
| 110 kDa | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 2081 | |
| Human, Mouse, Rat, Pig, Drosophila, Goat, Mammalia, Monkey, Primate, Rabbit, Golden Syrian Hamster | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, In vitro assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated | |
| O75460 | |
| ERN1 | |
| This IRE1 alpha [p Ser724] antibody was raised against a synthetic peptide surrounding the phosphorylated serine 724 of the human IRE1 alpha protein. [Swiss-Prot #O75460] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB100-2323 IRE1 alpha [p Ser724] Antibody detects IRE-1 alpha when phosphorylated at Ser724 residue. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido