Learn More
Invitrogen™ IRAK-M Recombinant Rabbit Monoclonal Antibody (23GB875), Invitrogen™
GREENER_CHOICE
Código de producto. 30089268
Change view
Click to view available options
Cantidad:
20 μL
100 μL
Tamaño de la unidad:
100 microlitros
20 microlitros
30089268 100 μL 100 microlitros
30089252 20 μL 20 microlitros
2 options
Código de producto. 30089268
Marca: Invitrogen™ MA554018
Rabbit Recombinant Monoclonal Antibody
IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IRAK-M
Aplicaciones Flow Cytometry, Western Blot, Immunocytochemistry
Clasificación Recombinant Monoclonal
Clon 23GB875
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
génica IRAK3
N.º de referencia del gen Q9Y616
Alias de gen 4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M
Símbolos de los genes IRAK3
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human IRAK3 (550-596AA).
Método de purificación Affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 11213
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG