Descripción
Immunogen sequence: AIGLNNLDKA YVERYANTLL SVKLEVLSQG QDNLSWNEIQ NCIDMVNAQI QEENDRVVAV GYINEAIDEG N Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.
Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.
Especificaciones
Especificaciones
| Antígeno | IQGAP2 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.7 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | IQGAP2 |
| N.º de referencia del gen | Q13576 |
| Alias de gen | 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2 |
| Símbolos de los genes | IQGAP2 |
| Mostrar más |