missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQGAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA578262
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (87%), Rat (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
Especificaciones
| IQGAP1 | |
| Polyclonal | |
| Unconjugated | |
| IQGAP1 | |
| AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8826 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.16 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P46940 | |
| IQGAP1 | |
| Recombinant protein encompassing a sequence within the center region of human IQGAP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido