missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.05 M, Penta Chemicals

Código de producto. 30020489
Change view
Click to view available options
Cantidad:
1 L
Tamaño de la unidad:
1 litro
Código de producto. Cantidad unitSize
30020489 1 L 1 litro
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30020489

Marca: PENTA CHEMICALS 1758011000

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.

About us

The company PENTA was established in 1990 under the trade name Ing. Petr Švec - PENTA s.r.o.. From the very beginning, we have been a solely Czech company which is managed by the Švec family, including company founder Petr Švec.

We specialize in the manufacture and distribution of pure, laboratory, pharmaceutical and specialty chemicals, which we market under the trade name Penta. We manufacture and supply a wide range of organic and inorganic chemicals, in large volumes, with a quality ranging from technical, to pure, to A.G. and ACS.

High quality and flexibility

Thanks to the high quality of our products, and our ability to fulfil specific customer requirements, we rank among the leading suppliers of qualified chemicals in the domestic market, and more recently we have also expanded into the global market.

We manufacture the chemicals in our own modern complex, which fulfils the strictest quality and safety criteria. Our large warehouse facilities, including two automated warehouses, enable us to accomodate client requirements. We package the products in accordance with all legislative requirements, and can deliver them to the customer using our own ADR transport on request.

Environmental responsibility

Our priority is to supply the maximum quality of chemicals, reliability, and environmental awareness. Our efforts are evidenced by many awards and certificates, including ISO 9001, ISO 14001 and the Responsible Care certificate for responsible business in the chemical industry.

Especificaciones

Nombre del producto químico o material Iodine
CAS 7553-56-2
Concentración 0.05 M
Envase Glass Bottle
Cantidad 1 L
compliance-icons
Nombre del producto
  • Iodine ready to use volumetric solution c(I
Palabra de advertencia
  • Atención
Clasificación
  • Toxicidad específica para determinados órganos Categoría 2
Indicaciones de peligro
  • H373-Puede provocar daños en los órganos.
Consejos de prudencia
  • P314-Consultar a un médico en caso de malestar.
Otros peligros
  • MIXTURE LIST-Contém: potassium iodide, iodine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.