missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Involucrin Monoclonal Antibody (SY5)

Descripción
MA5-11803 targets Involucrin in ELISA, IF, IHC (P), IP, and WB applications and shows reactivity with Canine, Human, Non-human primate, Porcine, and Rat samples. The MA5-11803 immunogen is human keratinocytes' involucrin.
Involucrin, a component of the keratinocyte crosslinked envelope, is found in the cytoplasm and crosslinked to membrane proteins by transglutaminase. Involucrin is expressed in a range of stratified squamous epithelia, including the cornea. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures it is expressed by all cells that have left the basal layer. Involucrin expression is abnormal in squamous cell carcinomas and premalignant lesions and is reduced in severe dysplasias of the larynx and cervix. Its gene is mapped to 1q21, among calpactin I light chain, trichohyalin, profillaggrin, loricrin, and calcyclin.
Especificaciones
Especificaciones
| Antígeno | Involucrin |
| Aplicaciones | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Clasificación | Monoclonal |
| Clon | SY5 |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| génica | IVL |
| N.º de referencia del gen | P07476, P18174, P18175, P48998 |
| Alias de gen | 1110019C06Rik; involucrin; IVL |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?