Learn More
Invitrogen™ Interferon alpha-8 Recombinant Rabbit Monoclonal Antibody (003)

Descripción
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Interferon alpha-B/IFNA8.
Especificaciones
Especificaciones
| Antígeno | Interferon alpha-8 |
| Aplicaciones | ELISA |
| Clasificación | Recombinant Monoclonal |
| Clon | 003 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative |
| génica | IFNA8 |
| N.º de referencia del gen | P32881 |
| Alias de gen | IFNA8; IFN-alpha-8; IFN-alphaB; Interferon; interferon alpha 8; interferon alpha type 201; interferon alpha-8; Interferon alpha-B; interferon alpha-B'; interferon alpha-B2; interferon, alpha 8; leIF B; RP11-354P17.14-001 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.