missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Interferon alpha-2 Monoclonal Antibody (D4), Invitrogen™
Descripción
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
Especificaciones
Especificaciones
| Antígeno | Interferon alpha-2 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | D4 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | IFNA2 |
| N.º de referencia del gen | P01563 |
| Alias de gen | alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?