Learn More
Integrin beta 1/CD29 Antibody (419127) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB17783JF525
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Equine,Human,Porcine samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Especificaciones
| Integrin beta 1/CD29 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin β1/CD29, Gln21-Asp728 (predicted), Accession # P05556 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 3688 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 419127 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Equine, Human, Pig | |
| Purified |