Learn More
Integrin Alpha 3 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F9G4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66070-100UL
Descripción
The Integrin Alpha 3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Integrin Alpha 3. This antibody recognizes human, pig, mouse antigen. The Integrin Alpha 3 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Especificaciones
| Integrin Alpha 3 | |
| 3F9G4 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse | |
| 155 kDa | |
| IgG2a |
| Western Blot | |
| 1000 μg/mL | |
| CD49C, FLJ34631, FLJ34704, FRP 2, Galactoprotein B3, GAP B3, GAPB3, Integrin alpha 3, ITGA3, MSK18, VCA 2, VL3A, VLA 3 subunit alpha, VLA3a | |
| 100 μL | |
| 3675 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |