Learn More
Integrin Alpha 3 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F9G4, Proteintech
Mouse Monoclonal Antibody
Marca:  Proteintech CL488-66070-20UL
 Ver más versiones de este producto
Código de producto. 18014265
  • 140.00€ / 20 microlitros
Descripción
The Integrin Alpha 3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Integrin Alpha 3. This antibody recognizes human, pig, mouse antigen. The Integrin Alpha 3 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Especificaciones
Especificaciones
Integrin Alpha 3
3F9G4
CoraLite Plus 488
Mouse
Primary
Human, Pig, Mouse
155 kDa
IgG2a
Western Blot
1000 μg/mL
CD49C, FLJ34631, FLJ34704, FRP 2, Galactoprotein B3, GAP B3, GAPB3, Integrin alpha 3, ITGA3, MSK18, VCA 2, VL3A, VLA 3 subunit alpha, VLA3a
20 μL
3675
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.