missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSRR Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA549383
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
INSRR Polyclonal Antibody for Western Blot
INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity. Receptor with tyrosine-protein kinase activity. Functions as a pH sensing receptor which is activated by increased extracellular pH. Activates an intracellular signaling pathway that involves IRS1 and AKT1/PKB.
Especificaciones
| INSRR | |
| Polyclonal | |
| Unconjugated | |
| Insrr | |
| Insrr; insulin receptor related receptor; insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IRR; IR-related receptor; Sirr | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 60663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q64716 | |
| Insrr | |
| KLH conjugated synthetic peptide between 1265-1295 amino acids from the C-terminal region of Mouse Insrr. | |
| 400 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido