missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSIG-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-59687
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
INSIG-2 Polyclonal specifically detects INSIG-2 in Human, Rat samples. It is validated for Western Blot.
Especificaciones
| INSIG-2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| INSIG-2, INSIG2 membrane protein, insulin induced gene 2, insulin induced protein 2, insulin-induced gene 2 protein, MGC26273 | |
| Rabbit | |
| 25 kDa | |
| 100 μL | |
| Primary | |
| Zebrafish 87%. | |
| Human, Rat, Pig, Bovine, Canine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y5U4 | |
| INSIG2 | |
| Synthetic peptides corresponding to INSIG2(insulin induced gene 2) The peptide sequence was selected from the middle region of INSIG2. Peptide sequence WWVPPCCGTASAVIGLLYPCIDRHLGEPHKFKREWSSVMRCVAVFVGINH. | |
| Affinity purified | |
| RUO | |
| 51141 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido