Learn More
Elma™ Insert Covers
Descripción
- Elma ultrasonic units can be operated with or without a cover
- For more efficient operation, Elma offers various insert covers made of robust and easy-to-clean stainless steel
- Covers make it possible to clean objects made of different materials simultaneously – but separately
- If used correctly, all Elma inserts will help to prevent damage to the ultrasonic tank and thus extend the service life of the ultrasonic cleaner
- Allows cleaning with aggressive fluids
- Prolongs the service life of the unit
Especificaciones
Especificaciones
| Tipo | Lid |
| Para utilizar con (equipo) | Fisherbrand Select, Fisherbrand EASY, Elmasonic S, Elmasonic Select |
| Modelo | 80 |