Learn More
Influenza A H7 Hemagglutinin Antibody (InA414) [Janelia Fluor™ 635], Novus Biologicals™
Descripción
Influenza A H7 Hemagglutinin Monoclonal antibody specifically detects Influenza A H7 Hemagglutinin in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA,Direct ELISA
Especificaciones
Especificaciones
| Antígeno | Influenza A H7 Hemagglutinin |
| Aplicaciones | ELISA, Western Blot, Sandwich ELISA, Direct ELISA |
| Clasificación | Monoclonal |
| Clon | InA414 |
| Conjugado | Janelia Fluor 635 |
| Formulación | 50mM Sodium Borate |
| Especie del huésped | Mouse |
| Inmunógeno | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Netherlands/219/03 (strain H7N7). |
| Método de purificación | Protein A purified |
| Cantidad | 0.25 mL |
| Mostrar más |