Learn More
INA Mouse anti-Human, Unconjugated, Clone: 3C12A8, Proteintech
Código de producto. 16889565 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16889565 20 μL 20 microlitros
16879565 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16889565 Proveedor Proteintech N.º de proveedor 671201IG20UL
Mouse Monoclonal Antibody
The INA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INA. This antibody recognizes human, mouse, rat, pig antigen. The INA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
Antígeno INA
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Monoclonal
Clon 3C12A8
Concentración 1000 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 9118
Especies diana Human, Mouse, Rat, Pig
Isotype IgG2a