Learn More
importin alpha 5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16848184 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16848184 150 μL 150 microlitros
16858184 20 μL 20 microlitros
2 options
Código de producto. 16848184
Marca: Proteintech 181371AP150UL
Rabbit Polyclonal Antibody
The import of proteins into the nucleus is a process that involves at least 2 steps. The first is an energy-independent docking of the protein to the nuclear envelope and the second is an energy-dependent translocation through the nuclear pore complex. Imported proteins require a nuclear localization sequence (NLS) which generally consists of a short region of basic amino acids or 2 such regions spaced about 10 amino acids apart. Proteins involved in the first step of nuclear import have been identified in different systems. These include the Xenopus protein importin and its yeast homolog, SRP1 (a suppressor of certain temperature sensitive mutations of RNA polymerase I in Saccharomyces cerevisiae), which bind to the NLS. KPNA2 protein interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in V(D)J recombination.
Especificaciones
Antígeno importin alpha 5
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica KPNA1
N.º de referencia del gen P52294, P83953, Q60960
Alias de gen IMA5, Importin alpha 5, Importin subunit alpha 1, IPOA5, Karyopherin subunit alpha 1, KPNA1, Nucleoprotein interactor 1, RAG cohort protein 2, RCH2, SRP1, SRP1 beta
Símbolos de los genes Kpna1
Especie del huésped Rabbit
Inmunógeno importin alpha5 Fusion Protein Ag12751
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16646, 288064, 3836
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG