missing translation for 'onlineSavingsMsg'
Learn More

Imiquimod, MedChemExpress

Código de producto. 16401396
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 mg
200 mg
500 mg
Tamaño de la unidad:
100 miligramos
200 miligramos
500 miligramos
Product Code. Cantidad unitSize
16401396 200 mg 200 miligramos
16428156 100 mg 100 miligramos
16492046 500 mg 500 miligramos
3 options
This item is not returnable. View return policy

Product Code. 16401396

Brand: Medchem Express HYB0180

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and COVID-19.

Imiquimod (R 837), an immune response modifier, is a selective toll like receptor 7 (TLR7) agonist. Imiquimod exhibits antiviral and antitumor effects in vivo. Imiquimod can be used for the research of external genital, perianal warts, cancer and COVID-19.

Specifications

Nombre del producto químico o material Imiquimod
Forma física Powder
CAS 99011-02-6
Color Blanco
Cantidad 200 mg
Notas de grado de pureza Research
Fórmula molecular C14H16N4
Para utilizar con (aplicación) COVID-19-anti-virus
Sinónimo R 837
Información de solubilidad H2O : 2.64 mg/mL (10.99 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 2 mg/mL (8.32 mM; ultrasonic and warming and heat to 70°C)
SMILES NC1=NC2=CC=CC=C2C3=C1N=CN3CC(C)C
Peso molecular (g/mol) 240.3
Formula Weight (peso de la fórmula) 240.3
Porcentaje de pureza 98.08%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.