missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT11/LILRA5 Antibody (711828), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB6754L
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ILT11/LILRA5 Monoclonal antibody specifically detects ILT11/LILRA5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Especificaciones
| ILT11/LILRA5 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 353514 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 711828 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7 | |
| Chinese hamster ovary cell line CHO-derived recombinant human LILRA5/CD85f, Gly42-Arg268, Accession # A6NI73 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido