Learn More
IL5RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16800494 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16800494 150 μL 150 microlitros
16810494 20 μL 20 microlitros
2 options
Código de producto. 16800494
Marca: Proteintech 126551AP150UL
Rabbit Polyclonal Antibody
Interleukin 5 (IL-5) is a soluble T cell-derived factor, also known as T cellreplacing factor (TRF), that induces B cell and eosinophil growth and differentiation. IL-5 exerts its biological activity through the IL-5 receptor (IL-5R),which is composed of two chains: an alpha chain that binds IL-5 with low affinity and a beta chain that does not bind IL-5, but together with the IL-5 alpha chain,constitutes the high affinity IL-5 receptor. The cytoplasmic domain of both the alpha and beta chains is essential for signal transduction. Specifically, the membrane-proximal proline-rich sequence of the ytoplasmic domain of the IL-5R receptor alpha subunit, IL-5Ralpha, is critical for the IL-5 induced proliferative response, expression of nuclear proto-oncogenes and tyrosine phosphorylation of cellular proteins, such as JAK1 and JAK2.
Especificaciones
Antígeno IL5RA
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica IL5RA
N.º de referencia del gen P21183, Q01344
Alias de gen CD125, CDw125, HSIL5R3, IL 5 receptor subunit alpha, IL 5R alpha, IL 5R subunit alpha, IL 5RA, IL5R, IL5RA, interleukin 5 receptor, alpha
Símbolos de los genes Il5ra
Especie del huésped Rabbit
Inmunógeno IL5RA Fusion Protein Ag3358
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 114103, 16192, 3568
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG