missing translation for 'onlineSavingsMsg'
Learn More

IL33 Mouse anti-Human, Clone: 2A9C7, Proteintech

Código de producto. 16837925 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
16837925 150 μL 150 microlitros
16847925 20 μL 20 microlitros
2 options
Este artículo no se puede devolver. View return policy

Product Code. 16837925

Brand: Proteintech 662351IG150UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-33 (Interleukin-33) is a 270 amino acid, highly divergent protein belonging to the IL-1 family with an IL-1-like C-terminal domain. IL-33 is a dual function protein that may function both as a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. IL-33 binds to and signals through IL1RL1/ST2 and its stimulation recruits MYD88, IRAK1, IRAK4, and TRAF6. IL-33 activates NF-kappaB and MAP kinases, and drives production of TH2-associated cytokines from in vitro polarized TH2 cells. In vivo, IL-33 induces the expression of IL-4, IL-5, and IL-13 and leads to severe pathological changes in mucosal organs. IL-33 is proteolytically converted to a mature form by CASP1 and is highly expressed in high endothelial venules found in tonsils, Peyer′s patches and mesenteric lymph nodes and is almost undetectable in placenta. Prolonged IL-33 treatment of mice led to the development of eosinophilia, splenomegaly, and severe pathological changes in mucosal organs such as lungs, esophagus and small intestine. Recent experiments have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine and an intracellular nuclear factor with transcriptional regulatory properties. Despite its predicted molecular weight, IL-33 will often run at higher molecular weight in SDS-PAGE. Studies have shown that IL-33 can also co-localize with heterochromatin and possesses transcriptional repressor activities, indicating that IL-33 may function as both a proinflammatory cytokine, and an intracellular nuclear factor with transcriptional regulatory properties.

Specifications

Antígeno IL33
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Clon 2A9C7
Concentración 1.36 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica IL33
N.º de referencia del gen O95760
Alias de gen C9orf26, DVS27, IL 1F11, IL 33, IL1F11, IL33, Interleukin 1 family member 11, interleukin 33, NF HEV, NFEHEV, NFHEV, RP11 575C20.2
Símbolos de los genes IL33
Especie del huésped Mouse
Inmunógeno IL33 Fusion Protein Ag21430
Método de purificación Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 90865
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.