Learn More
IL22RA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16861694 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16861694 20 μL 20 microlitros
16851694 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16861694 Proveedor Proteintech N.º de proveedor 134621AP20UL
Rabbit Polyclonal Antibody
A novel cytokine, designated IL-TIF for IL-10 related T cell-derived inducible factor and IL-22, was recently identified. The receptor for IL-22 (IL-22R, also termed CRF2-9 and IL-TIF-R1 chain) is a new member of the class II cytokine receptor family. IL-22R forms a complex with IL-10 receptor beta chain and mediates IL-22 signaling. IL-22 and its receptor activate JAK-STAT signaling pathway. IL22R is expressed in normal liver and kidney and their cell lines HepG2 and TK-10. A soluble form of IL-22 receptor, also termed IL-22 binding protein (IL-22BP) and IL-22 receptor-alpha 2 (IL-22RA2), was identified very recently. IL-22BP prevents binding of IL-22 to the functional cell surface IL-22R complex and neutralizes IL-22 activity. LPS induces IL-22 expression, which indicates the role of IL-22 in inflammatory response.
Especificaciones
Antígeno IL22RA1
Aplicaciones Flow Cytometry, Immunofluorescence, Immunocytochemistry, Western Blot
Clasificación Polyclonal
Concentración 0.53 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica IL22RA1
N.º de referencia del gen Q80XZ4, Q8N6P7
Alias de gen CRF2 9, IL 22 receptor subunit alpha 1, IL 22R alpha 1, IL 22RA1, IL22R, IL22R1, IL22RA1, ZcytoR11
Símbolos de los genes Il22ra1
Especie del huésped Rabbit
Inmunógeno IL22RA1 Fusion Protein Ag4056
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 230828, 362629, 58985
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG