Learn More
PeproTech IL17B Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Descripción
AA Sequence of recombinant protein: MQPRSPKSKR KGQGRPGPLA PGPHQVPLDL VSRMKPYARM EEYERNIEEM VAQLRNSSEL AQRKCEVNLQ LWMSNKRSLS PWGYSINHDP SRIPVDLPEA RCLCLGCVNP FTMQEDRSMV SVPVFSQVPV RRRLCPPPPR TGPCRQRAVM ETIAVGCTCI F. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17B. Anti-Human IL-17B-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIL-17B by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-17B (500-P248) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIL-17B. Western Blot: To detect hIL-17B by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hIL-17B is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P248BT-1MG will be provided as 2 x 500 μg
Especificaciones
Especificaciones
| Antígeno | IL17B |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1-1.0 mg/mL |
| Conjugado | Biotin |
| Formulación | PBS with no preservative |
| génica | IL17B |
| N.º de referencia del gen | Q9UHF5 |
| Alias de gen | 1110006O16Rik; 1700006N07Rik; Cytokine CX1; Cytokine Zcyto7; Cytokine-like protein ZCYTO7; gene for cytokine-like protein; IL17B; IL-17B; IL20; IL-20; Interleukin; interleukin 17B; interleukin 20; interleukin-17 beta; interleukin-17B; Interleukin20; Interleukin-20; neuronal interleukin-17 related factor; neuronal interleukin-17-related factor; NIRF; UNQ516/PRO1031; ZCYTO7 |
| Símbolos de los genes | IL17B |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.