missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-9 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. 17804863
Change view
Click to view available options
Cantidad:
2 x 500μg
25 μg
50 μg
500 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
50 microgramos
500 microgramos
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17804863 50 μg 50 microgramos
17875673 25 μg 25 microgramos
17835733 2 x 500μg 1 miligramo
30054739 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17804863 Proveedor PeproTech N.º de proveedor 500P29BT50UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-9. Anti-Human IL-9-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-9 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-9 (500-P29) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-9. Western Blot: To detect Human IL-9 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human IL-9 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P29BT-1MG will be provided as 2 x 500 μg

Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IL-9
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Biotin
Formulación PBS with no preservative
génica Il9
N.º de referencia del gen P15248
Alias de gen cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40
Símbolos de los genes Il9
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human IL-9.
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3578
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.