Learn More
MedChemExpress IL-8/CXCL8 Protein, Human (CHO)
Especificaciones
Especificaciones
| Contenido y almacenamiento | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| Cantidad | 10 μg |
| Condiciones de envío | Room temperature |
| Peso molecular | Approximately 9 kDa |
| Sinónimo | rHuIL-8/CXCL8∣ C-X-C motif chemokine 8∣ NAP-1∣ IL8∣ GCP-1 |