missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-6 Mouse anti-Human, Clone: OTI1D9, TrueMAB™, Origene
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antígeno | IL-6 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | OTI1D9 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | Il6 |
| N.º de referencia del gen | P05231 |
| Alias de gen | B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?