missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-4 Monoclonal Antibody (24G2), Biotin

Código de producto. 11894331
Change view
Click to view available options
Cantidad:
250 μg
Tamaño de la unidad:
500 microgramos
Código de producto. Cantidad unitSize
11894331 250 μg 500 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11894331

Marca: Invitrogen™ MM450DB

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

MM450DB targets IL-4 in ELISA, and WB applications and shows reactivity with mouse samples. The MM450DB immunogen is recombinant mouse IL-4. MM450DB detects IL-4 which has a predicted molecular weight of approximately 14 kDa. The MM450DB mouse IL4 antibody (biotinylated conjugate of clone 24G2) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody MM450C (clone 1D11). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Biotinylated antibody MM450DB (clone 24G2) and antibody MM450C (clone 1D11) have successfully been used in combination with recombinant mouse IL4 protein SMIL4 in ELISA applications. This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IL-4
Aplicaciones ELISA, Western Blot
Clasificación Monoclonal
Clon 24G2
Concentración 0.5 mg/mL
Conjugado Biotin
Formulación PBS with 4% BSA and proprietary Preservative
génica Il4
N.º de referencia del gen P07750
Alias de gen B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Símbolos de los genes Il4
Especie del huésped Rat
Inmunógeno Recombinant mouse IL-4.
Método de purificación Protein A
Cantidad 250 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 16189
Especies diana Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.