missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-3 Monoclonal Antibody (4815)
GREENER_CHOICE

Código de producto. 15583382
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
500 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15583382 500 μg 500 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15583382 Proveedor Invitrogen™ N.º de proveedor MA523695

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

In sandwich ELISAs, no significant cross-reactivity or interference with recombinant human (rh) IL-1 alpha, rhIL-1 beta, recombinant mouse (rm) IL-1 beta, rhIL-2, rhIL-3 R alpha, rhIL-3 R beta, rmIL-3, rhIL-4, rmIL-4, rmIL-5, rhIL-6, rmIL-6, rhIL-7, rmIL-7, rhIL-8, bovine FGF acidic, bovine FGF basic, rhG-CSF, rhGM-CSF, rmGM-CSF, rhLIF, hPDGF, porcine PDGF, human TGF- beta 1, porcine TGF- beta 1, rhTGF- beta 1, porcine TGF- beta 2, porcine TGF- beta 1.2, rhTNF-alpha, or rhTNF-beta was observed. This antibody does not neutralize the biological activity of rmIL-3. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IL-3
Aplicaciones ELISA, Neutralization
Clasificación Monoclonal
Clon 4815
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 5% trehalose and No Preservative
génica IL3
N.º de referencia del gen P08700
Alias de gen BP/BPA; BPA; Colony stimulating factor multiple; colony-stimulating factor, multiple; Csfmu; Eo-CSF; HCGF; hematopoietic growth factor; Il3; Il-3; ILN; Interleukin; interleukin 3; Interleukin 3 (colony stimulating factor; Interleukin3; interleukin-3; Mast cell growth factor; mast-cell growth factor; MCGF; MGC79398; MGC79399; MGF; Multi CSF; MULTI-CSF; Multilineage colony stimulating factor; multilineage-colony-stimulating factor; multiple); multipotential colony-stimulating factor; P cell stimulating factor; P-cell stimulating factor; P-cell-stimulating factor; PCSA; PSF
Símbolos de los genes IL3
Especie del huésped Mouse
Inmunógeno E. coli-derived recombinant human IL-3 Ala20-Phe152.
Método de purificación Protein A/G
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3562
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.