missing translation for 'onlineSavingsMsg'
Learn More

IL-23A Mouse anti-Human, Unconjugated, Clone: 1D5D3, Proteintech

Código de producto. 16807855 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
16807855 20 μL 20 microlitros
16897845 150 μL 150 microlitros
2 options
This item is not returnable. View return policy

Product Code. 16807855

Brand: Proteintech 661961IG20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The IL-23A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-23A. This antibody recognizes human antigen. The IL-23A antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Antígeno IL-23A
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Monoclonal
Clon 1D5D3
Concentración 747 μg/mL
Conjugado Unconjugated
Especie del huésped Mouse
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 51561
Especies diana Human
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.