missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-19 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Código de producto. 17875803
Change view
Click to view available options
Cantidad:
2 x 500 μg
25 μg
50 μg
500 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
17875803 50 μg 50 microgramos
17834943 25 μg 25 microgramos
17855133 2 x 500 μg 1 miligramo
30054785 500 μg 500 microgramos
4 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17875803

Marca: PeproTech 500P189BT50UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MLRRCLISTD MHHIEESFQE IKRAIQAKDT FPNVTILSTL ETLQIIKPLD VCCVTKNLLA FYVDRVFKDH QEPNPKILRK ISSIANSFLY MQKTLRQCQE QRQCHCRQEA TNATRVIHDN YDQLEVHAAA IKSLGELDVF LAWINKNHEV MSSA. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-19. Anti-Human IL-19-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hIL-19 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-19 (500-P189) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hIL-19. Western Blot: To detect Human IL-19 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-19 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P189BT-1MG will be provided as 2 x 500 μg

The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IL-19
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Concentración 0.1-1.0 mg/mL
Conjugado Biotin
Formulación PBS with no preservative
génica IL19
N.º de referencia del gen Q9UHD0
Alias de gen il 19; IL-10C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG.1; ZMDA1
Símbolos de los genes IL19
Especie del huésped Rabbit
Inmunógeno E.coli-derived Recombinant Human IL-19.
Método de purificación Antigen affinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 29949
Especies diana Human
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Lyophilized
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.