Learn More
IL-17E/IL-25 Antibody (68C1039.2) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NBP2-33223
Código de producto. 18199483
  • ONLINE EXCLUSIVE PRICE
    584.00€
    552.30€ / 0.10 miligramo
    Ahorro 31.70€
    5% Descuento
Descripción
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
IL-17E/IL-25
Monoclonal
1.0 mg/mL
Western Blot 2-4 ug/ml, Immunohistochemistry 1:20-1:1000, Immunohistochemistry-Paraffin 1:10-1:500. Use reported in scientific literature (Terrier B et al)
IL-17E, IL17Einterleukin-17E, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25
Mouse
20 kDa
0.1 mg
Cytokine Research
64806
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
68C1039.2
Unconjugated
Q9H293
IL25
Amino acids 115-132 of human IL-17E (isoform CRA_a) were used as immunogen for this antibody.
Protein G purified
RUO
Primary
Human, Mouse
Purified
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones