Learn More
Invitrogen™ Human IL-13 ProQuantum Immunoassay Kit
Descripción
The ProQuantum Human IL-13 Immunoassay Kit is designed to provide highly sensitive quantitative measurements of Human IL-13 protein in small sample volumes. Utilizing proximity ligation assay (PLA) technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive Cloud-based software for analytics enables sample-to-answer in just 2 hours. High sensitivity-detect low levels of protein with greater sensitivity than traditional methods Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range Small sample consumption-use 2-5 μL of sample (compared to 75 μL for triplicate wells with other methods) Fast, easy workflow-2 hours from sample to answer, with no wash steps No proprietary instrument to purchase-runs on any real-time PCR instrument ProQuantum immunoassays utilize a matched pair of target-specific antibodies, each conjugated to a DNA oligonucleotide. During antibody-analyte binding, the two DNA oligos are brought into close proximity, which allows for ligation of the two strands and subsequent creation of a template strand for amplification. This platform leverages the sensitivity and large dynamic range of Applied Biosystems TaqMan real-time...
- Kit de inmunoensayo ProQuantum
- Para ELISA
Especificaciones
Especificaciones
| Número de acceso | P35225 |
| Intervalo del ensayo | 0.0128-5000 pg/mL |
| Sensibilidad del ensayo | <0.01 pg/mL |
| Contenido y almacenamiento | Contiene un tampón de dilución de ensayo, un conjugado de anticuerpos A, un conjugado de anticuerpos B, un tampón de dilución de conjugado de anticuerpos, un patrón, una mezcla maestra y una ligasa. Almacenar de -5 °C a -30 °C. |
| Conjugado | FAM |
| Formato del kit de inmunoensayo | Proximity-based Immunoassay |
| Tipo de producto | ProQuantum |
| Tipo de muestra | Plasma, Serum, Supernatant |
| Especies diana | Human |
| Para utilizar con (equipo) | qPCR Instruments |
| Mostrar más |