missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IHOG Monoclonal Antibody (3G8)
GREENER_CHOICE

Código de producto. 11365613
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11365613 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11365613 Proveedor Invitrogen™ N.º de proveedor MA515648

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-15648 targets IHOG in indirect ELISA, WB applications and shows reactivity with Drosophila samples. The MA5-15648 immunogen is purified recombinant fragment of human IHOG expressed in E. Coli. MA5-15648 detects IHOG which has a predicted molecular weight of approximately 98kDa.

The ihog gene (interference hedgehog), identified by RNA interference in Drosophila cultured cells, encodes a type 1 membrane protein shown here to bind and to mediate response to the active Hedgehog (Hh) protein signal. ihog mutations produce defects characteristic of Hh signaling loss in embryos and imaginal discs, and epistasis analysis places ihog action at or upstream of the negatively acting receptor component, Patched (Ptc). The first of two extracellular fibronectin type III (FNIII) domains of the Ihog protein mediates a specific interaction with Hh protein in vitro, but the second FNIII domain is additionally required for in vivo signaling activity and for Ihog-enhanced binding of Hh protein to cells coexpressing Ptc. Other members of the Ihog family, including Drosophila Boi and mammalian CDO and BOC, also interact with Hh ligands via a specific FNIII domain, thus identifying an evolutionarily conserved family of membrane proteins that function in Hh signal response.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno IHOG
Aplicaciones ELISA, Western Blot
Clasificación Monoclonal
Clon 3G8
Concentración Conc. Not Determined
Conjugado Unconjugated
Formulación ascites with 0.03% sodium azide
génica ihog
N.º de referencia del gen Q9VM64
Alias de gen CG9211; CG9211-PA; CT26314; Dmel\ ihog; Dmel\CG9211; Dmel_CG9211; GH03927p; Ihog; ihog-PA; interference hedgehog
Símbolos de los genes ihog
Especie del huésped Mouse
Inmunógeno Purified recombinant fragment of human IHOG expressed in E. Coli.
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 33972
Especies diana Human, Drosophila
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.