Learn More
IgG4 Mouse anti-Human, CoraLite 488, Clone: 1A1A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66408-100UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Human IgG4 antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Human IgG4. This antibody recognizes human antigen. The Human IgG4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Especificaciones
| IgG4 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 3503 | |
| IgG1 |
| Immunofluorescence | |
| 1A1A2 | |
| Mouse | |
| Primary | |
| Human |