missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2b Mouse Anti-Rat, Brilliant Violet 711, Clone: G15-337, BD Optibuild™
Descripción
The G15-337 monoclonal antibody specifically recognizes rat IgG2b. It does not react with other Ig isotypes. A suspension of pooled rat Ig was used as the source of immunogen.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Especificaciones
Especificaciones
| Antígeno | IgG2b |
| Clasificación | Monoclonal |
| Clon | G15-337 |
| Concentración | 0.2mg/mL |
| Conjugado | Brilliant Violet 711 |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Alias de gen | Ig gamma-2B, Immunoglobulin G2b, Cγ2b |
| Especie del huésped | Mouse |
| Inmunógeno | Pooled rat Ig |
| Método de purificación | Affinity Purified |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?