missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG2 Mouse anti-Mouse, BV750, Clone: G192-3, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 747088
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
IgG2 Monoclonal antibody specifically detects IgG2 in Mouse samples. It is validated for Flow Cytometry
Especificaciones
| IgG2 | |
| Monoclonal | |
| Brilliant Violet 750 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| G192-3 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Pooled Armenian and Syrian Hamster IgG mAb | |
| RUO | |
| Mouse | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido