missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1/IgG2a Mouse Anti-Rat, Brilliant Violet 650, Clone: G28-5, BD Optibuild™
Mouse Monoclonal Antibody
Marca: BD Biosciences 743852
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The G28-5 monoclonal antibody reacts specifically with rat IgG1 and IgG2a. It does not react with other Ig isotypes. A suspension of pooled rat IgG1 was used as the source of immunogen.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Especificaciones
| IgG1/IgG2a | |
| G28-5 | |
| Brilliant Violet 650 | |
| IgG1/IgG2a, Ig gamma-1/2a, Immunoglobulin gamma 1/2a | |
| Pooled Rat IgG | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido